Lithium aluminum hydride 2 mol/l THF, Sigma-Aldrich®

Supplier: Merck

Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate

MQ200

593702-100ML
SIAL593702-100MLEA 77 EUR
SIAL593702-100ML
Lithium aluminum hydride 2 mol/l THF, Sigma-Aldrich®
Lithium aluminum hydride
Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1411
ADR: 4.3 I
Merck Index: 13,00348

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR