N-Lauroylsarcosine sodium salt, Sigma-Aldrich®

Supplier: Merck
Danger

61743-25G
SIAL61743-25GEA 402 EUR
SIAL61743-25G
N-Lauroylsarcosine sodium salt, Sigma-Aldrich®
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
Melting Pt: 45…47 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,II
Merck Index: 12,04379
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR