Lithium acetate dihydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Acetic acid lithium salt

62393-100G-F 62393-500G-F
SIAL62393-100G-FEA 119 EUR
SIAL62393-100G-F SIAL62393-500G-F
Lithium acetate dihydrate, Sigma-Aldrich®
Lithium acetate dihydrate
Formula: H₃CCOOLi·2H₂O
Melting Pt: 291 °C
Density: 1.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR