Methyl malonyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methyl 3-chloro-3-oxopropanoate

M-Clarity™ quality level = MQ200

63406-10ML 63406-50ML
SIAL63406-10MLEA 170 EUR
SIAL63406-10ML SIAL63406-50ML
Methyl malonyl chloride, Sigma-Aldrich®
Methyl malonyl chloride
Formula: C₄H₅ClO₃
MDL Number: MFCD00013657
CAS Number: 37517-81-0
EINECS: 253-540-6
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR