Silicon dioxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

M-Clarity™ quality level = MQ100

634867-5EA
SIAL634867-5EAEA 213 EUR
SIAL634867-5EA
Silicon dioxide, Sigma-Aldrich®
Silicium dioxide
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1719
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR