3-Methylphenethyl bromide, Sigma-Aldrich®

Supplier: Merck

692018-5G
SIAL692018-5GEA 127 EUR
SIAL692018-5G
3-Methylphenethyl bromide, Sigma-Aldrich®
3-Methylphenethyl bromide
Formula: C₉H₁₁Br
CAS Number: 16799-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR