Sodium-tert-butanolate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Sodium tert-butoxide, Sodium t-butoxide, Sodium Tert-Butylate

703788-25G 703788-5G
SIAL703788-25GEA 161 EUR
SIAL703788-25G SIAL703788-5G
Sodium-tert-butanolate, Sigma-Aldrich®
Sodium tert-butanolate
Formula: C₄H₉NaO
MDL Number: MFCD00040575
CAS Number: 865-48-5
EINECS: 212-741-9
UN: 3206
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR