Potassium ethyltrifluoroborate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Potassium ethyltrifluoroboranuide

715212-1G
SIAL715212-1GEA 139 EUR
SIAL715212-1G
Potassium ethyltrifluoroborate, Sigma-Aldrich®
Potassium ethyltrifluoroborate
Formula: C₂H₅BF₃K
MDL Number: MFCD04112713
CAS Number: 44248-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR