Phthaloyl dichloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Phthaloyl chloride, o-Phthaloyl dichloride

79830-100ML 79830-500ML
SIAL79830-100MLEA 40 EUR
SIAL79830-100ML SIAL79830-500ML
Phthaloyl dichloride, Sigma-Aldrich®
Phthaloyl dichloride
Formula: C₈H₄Cl₂O₂
MDL Number: MFCD00000666
CAS Number: 88-95-9
EINECS: 201-869-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR