1,2-Dichloropropane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Propylene dichloride

M-Clarity™ quality level = MQ100

82270-1L 82270-250ML
SIAL82270-1LEA 125 EUR
SIAL82270-1L SIAL82270-250ML
1,2-Dichloropropane, Sigma-Aldrich®
1,2-Dichloropropane
Formula: ClH₂CCH₂CH₂Cl
Boiling Pt: 96 °C (1013 hPa)
Melting Pt: –100 °C
Density: 1.158 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000868
CAS Number: 78-87-5
EINECS: 201-152-2
UN: 1279
ADR: 3,II
Merck Index: 12,08039

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR