Tetrabutylammonium iodide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

86890-100G 86890-25G 86890-500G
SIAL86890-100GEA 62 EUR
SIAL86890-100G SIAL86890-25G SIAL86890-500G
Tetrabutylammonium iodide, Sigma-Aldrich®
Tetrabutylammonium iodide
Formula: C₁₆H₃₆IN
Melting Pt: 143…146 °C
Density: 1.0027 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011636
CAS Number: 311-28-4
EINECS: 206-220-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR