Sodium alginate (from Macrocystis pyrifera (kelp)), powder, Bioreagent plant cell culture tested, Sigma-Aldrich®

Supplier: Merck

Synonyms: Alginic acid sodium salt, Algin

M-Clarity™ quality level = MQ200

A0682-100G
SIALA0682-100GEA 104 EUR
SIALA0682-100G
Sodium alginate (from Macrocystis pyrifera (kelp)), powder, Bioreagent plant cell culture tested, Sigma-Aldrich®
Sodium alginate
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR