Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate

M-Clarity™ quality level = MQ200

A26209-10G A26209-1G A26209-5G
SIALA26209-10GEA 352 EUR
SIALA26209-10G SIALA26209-1G SIALA26209-5G
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Adenosine 5'-triphosphate disodium salt (ATP disodium salt) hydrate
Formula: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MDL Number: MFCD00150755
CAS Number: 34369-07-8
EINECS: 213-579-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR