Adenosine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 9-β-D-Ribofuranosyladenine, (2R,3R,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, Adenine-9-β-D-ribofuranoside, Adenine riboside

M-Clarity™ quality level = MQ200

A4036-25G A4036-5G
SIALA4036-25GEA 164 EUR
SIALA4036-25G SIALA4036-5G
Adenosine, Sigma-Aldrich®
Adenosine
Formula: C₁₀H₁₃N₅O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00005752
CAS Number: 58-61-7
EINECS: 200-389-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR