Bromophenol blue, Sigma-Aldrich®

Supplier: Merck

Synonyms: Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein

M-Clarity™ quality level = MQ200

B0126-25G
SIALB0126-25GEA 79.6 EUR
SIALB0126-25G
Bromophenol blue, Sigma-Aldrich®
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
Melting Pt: 273 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2
Merck Index: 13,01429

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR