Chloroacetic acid, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

C19627-1KG C19627-25G
SIALC19627-1KGEA 81.7 EUR
SIALC19627-1KG SIALC19627-25G
Chloroacetic acid, Sigma-Aldrich®
Chloroacetic acid
Formula: ClCH₂COOH
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: 56…57 °C
Density: 1.4043 g/cm³ (20 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1,II
Merck Index: 13,02129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2516e0c26c6c26f4f6f3656bc5b4256e7cf08207&vid=9c9b7ad094469d9089eb2f0ce88275d1423c2b46&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR