3-Chlorobenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ100

C23403-100G C23403-25G
SIALC23403-100GEA 109 EUR
SIALC23403-100G SIALC23403-25G
3-Chlorobenzaldehyde, Sigma-Aldrich®
3-Chlorobenzaldehyde
Formula: C₇H₅ClO
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 17…18 °C
Density: 1.241 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR