3,5-Dimethylbenzoic acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

D149608-25G
SIALD149608-25GEA 86.2 EUR
SIALD149608-25G
3,5-Dimethylbenzoic acid, Sigma-Aldrich®
3,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
Boiling Pt: 278 °C (1013 hPa)
Melting Pt: 166…169 °C
Density: 1.211 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR