2,3-Dimethylbutane, Sigma-Aldrich®

Supplier: Merck
Danger

D151602-500ML D151602-100ML
SIALD151602-500MLEA 269 EUR
SIALD151602-500ML SIALD151602-100ML
2,3-Dimethylbutane, Sigma-Aldrich®
2,3-Dimethylbutane
Formula: C₆H₁₄
Boiling Pt: 58 °C (1013 hPa)
Melting Pt: –128.5 °C
Density: 0.6615 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008925
CAS Number: 79-29-8
EINECS: 201-193-6
UN: 2457
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR