Ammonium iron(II) sulphate hexahydrate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

F1543-1KG F1543-500G
SIALF1543-1KGEA 361 EUR
SIALF1543-1KG SIALF1543-500G
Ammonium iron(II) sulphate hexahydrate, Sigma-Aldrich®
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
Melting Pt: 100 °C
Density: 1.864 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-151-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR