Iron(III) nitrate nonahydrate, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

F8508-100G
SIALF8508-100GEA 48.1 EUR
SIALF8508-100G
Iron(III) nitrate nonahydrate, Sigma-Aldrich®
Iron (III) nitrate nonahydrate
Formula: Fe(NO₃)₃·9H₂O
Boiling Pt: 125 °C (1013 hPa)
Melting Pt: 47 °C
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1,III
Merck Index: 13,04057

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR