(±)-4-Hydroxy-3-methoxymandelic acid, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

H0131-1G H0131-250MG H0131-100MG H0131-500MG H0131-5G
SIALH0131-1GEA 185 EUR
SIALH0131-1G SIALH0131-250MG SIALH0131-100MG SIALH0131-500MG SIALH0131-5G
(±)-4-Hydroxy-3-methoxymandelic acid, Sigma-Aldrich®
(±)-4-Hydroxy-3-methoxymandelic acid
Formula: C₉H₁₀O₅
Boiling Pt: 421 °C (1013 hPa)
Melting Pt: 132…134 °C
Density: 1.44 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004235
CAS Number: 55-10-7
EINECS: 200-224-0
Merck Index: 12,10070

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR