HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid), Sigma-Aldrich®

Supplier: Merck

Synonyms: HEPES free acid, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid), 4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid, N-2-Hydroxyethylpiperazine-N′-2-ethanesulphonic acid, 2-(4-(2-Hydroxyethyl)piperazin-1-yl)ethanesulphonic acid

M-Clarity™ quality level = MQ400

H4034-100G H4034-1KG H4034-25G H4034-25KG H4034-500G
SIALH4034-100GEA 182 EUR
SIALH4034-100G SIALH4034-1KG SIALH4034-25G SIALH4034-25KG SIALH4034-500G
HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid), Sigma-Aldrich®
HEPES (4-(2-Hydroxyethyl)piperazin-1-ylethanesulphonic acid)
Formula: C₈H₁₈N₂O₄S
Melting Pt: 210…215 °C
Density: 1.448 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006158
CAS Number: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cdc09c866676149941b93b2cac8d453175059a2&vid=0b09e5b84ea34a806443c3071fad63e15e4e3433&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR