3-(Hydroxymethyl)indole, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Indolmethanol, 3-Indolcarbinol, (1H-Indol-3-yl)methanol, Indole-3-methanol, (Indole-3-yl)methanol, 3-Indolemethanol, Indole-3-carbinol

M-Clarity™ quality level = MQ100

I7256-1G I7256-25G I7256-5G
SIALI7256-1GEA 31.3 EUR
SIALI7256-1G SIALI7256-25G SIALI7256-5G
3-(Hydroxymethyl)indole, Sigma-Aldrich®
3-(Hydroxymethyl)indole
Formula: C₉H₉NO
MDL Number: MFCD00005632
CAS Number: 700-06-1
EINECS: 211-836-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR