Isoamyl alcohol, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

I9392-25ML I9392-4X25ML I9392-500ML
SIALI9392-25MLEA 50.8 EUR
SIALI9392-25ML SIALI9392-4X25ML SIALI9392-500ML
Isoamyl alcohol, Sigma-Aldrich®
Isoamyl alcohol
Formula: (CH₃)₂CHCH₂CH₂OH
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.812 g/cm³ (20 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00002934
CAS Number: 123-51-3
EINECS: 204-633-5
UN: 1105
ADR: 3,III
Merck Index: 13,05214
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd7b89cc1890b5d4b2083195c30c0d1047c32e9a&vid=170c60e26c7b4edab6e0686f91d04e676da16e6d&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR