Lithium dodecyl sulphate, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt

M-Clarity™ quality level = MQ200

L5901-25G L5901-5G
SIALL5901-25GEA 378 EUR
SIALL5901-25G SIALL5901-5G
Lithium dodecyl sulphate, Sigma-Aldrich®
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR