Myrcene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 7-Methyl-3-methyleneocta-1,6-diene

M-Clarity™ quality level = MQ200

M100005-100ML M100005-500ML M100005-5ML
SIALM100005-100MLEA 37.4 EUR
SIALM100005-100ML SIALM100005-500ML SIALM100005-5ML
Myrcene, Sigma-Aldrich®
Myrcene
Formula: C₁₀H₁₆
MDL Number: MFCD00008908
CAS Number: 123-35-3
EINECS: 204-622-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR