Sodium molybdate(VI) dihydrate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

M-Clarity™ quality level = MQ200

M1651-100G
SIALM1651-100GEA 91.2 EUR
SIALM1651-100G
Sodium molybdate(VI) dihydrate, Sigma-Aldrich®
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
Melting Pt: 687 °C
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR