Hydroquinone monomethyl ether, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

M18655-1KG M18655-100G M18655-500G M18655-5G
SIALM18655-1KGEA 113 EUR
SIALM18655-1KG SIALM18655-100G SIALM18655-500G SIALM18655-5G
Hydroquinone monomethyl ether, Sigma-Aldrich®
Hydroquinone monomethyl ether
Formula: C₇H₈O₂
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 55…57 °C
Density: 1.55 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR