Methyl chloroformate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

M35304-100G M35304-500G M35304-5G
SIALM35304-100GEA 51.5 EUR
SIALM35304-100G SIALM35304-500G SIALM35304-5G
Methyl chloroformate, Sigma-Aldrich®
Methyl chloroformate
Formula: C₂H₃ClO₂
Boiling Pt: 71…72 °C (1013 hPa)
Melting Pt: –61 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00000639
CAS Number: 79-22-1
EINECS: 201-187-3
UN: 1238
ADR: 6.1,I
Merck Index: 12,06123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR