(±)-2-Methylpiperazine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Methylpiperazine

M-Clarity™ quality level = MQ100

M72404-100G
SIALM72404-100GEA 100 EUR
SIALM72404-100G
(±)-2-Methylpiperazine, Sigma-Aldrich®
(±)-2-Methylpiperazine
Formula: C₅H₁₂N₂
Boiling Pt: 155 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00005954
CAS Number: 109-07-9
EINECS: 203-644-2
UN: 2925
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR