1-Naphthyl isothiocyanate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

N4525-10G
SIALN4525-10GEA 145 EUR
SIALN4525-10G
1-Naphthyl isothiocyanate, Sigma-Aldrich®
1-Naphthyl isothiocyanate
Formula: C₁₁H₇NS
Boiling Pt: 100 °C (0.2 mmHg)
Melting Pt: 59 °C
Density: 1.81 g/cm³ (20 °C)
MDL Number: MFCD00003882
CAS Number: 551-06-4
EINECS: 208-990-8
UN: 2923
ADR: 8,III
Merck Index: 12,06498

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR