β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®

Supplier: Merck

N9410-15VL
SIALN9410-15VLEA 383 EUR
SIALN9410-15VL
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na₂, reduced form), Sigma-Aldrich®
β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form)
Formula: C₂₁H₂₇N₇Na₂O₁₄P₂
MDL Number: MFCD00036200
CAS Number: 606-68-8
EINECS: 210-123-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR