Penicillin G potassium salt, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 6-(2-Phenylacetamido)penicillanic acid potassium salt, Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin G potassium, Potassium [2S-(2α,5α,6β)]-3,3-dimethyl-7-oxo-6-(phenylacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Benzylpenicillin potassium salt

M-Clarity™ quality level = MQ200

P8721-100MU
SIALP8721-100MUEA 373 EUR
SIALP8721-100MU
Penicillin G potassium salt, Sigma-Aldrich®
Penicillin G potassium salt
Formula: C₁₆H₁₇KN₂O₄S
MW: 372.49 g/mol
Melting Pt: 214…217 °C
Density: 1.359 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00036193
CAS Number: 113-98-4
EINECS: 204-038-0
Merck Index: 12,07225

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR