o-Phenylenediamine dihydrochloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Benzene-1,2-diamine dihydrochloride, 1,2-Phenylenediammonium dichloride, 1,2-Diaminobenzene dihydrochloride, 1,2-Phenylenediamine dihydrochloride

M-Clarity™ quality level = MQ300

P8787-100TAB
SIALP8787-100TABEA 609 EUR
SIALP8787-100TAB
o-Phenylenediamine dihydrochloride, Sigma-Aldrich®
o-Phenylenediamine dihydrochloride
Formula: C₆H₁₀Cl₂N₂
Melting Pt: 205 °C
MDL Number: MFCD00012966
CAS Number: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR