Sodium peroxodisulphate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Sodium peroxidisulphate, Sodium persulphate, Disodium peroxodisulphate, Peroxydisulphuric acid disodium salt

M-Clarity™ quality level = MQ100

S6172-1KG S6172-250G
SIALS6172-1KGEA 279 EUR
SIALS6172-1KG SIALS6172-250G
Sodium peroxodisulphate, Sigma-Aldrich®
Sodium peroxodisulphate
Formula: Na₂S₂O₈
Melting Pt: 100 °C
Density: 2.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003501
CAS Number: 7775-27-1
EINECS: 231-892-1
UN: 1505
ADR: 5.1,III
Merck Index: 13,08729

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR