TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: TES Buffer, N-Tris(hydroxymethyl)methyl-2-aminoethanesulphonic acid

M-Clarity™ quality level = MQ200

T1375-100G T1375-1KG T1375-25G T1375-500G T1375-5KG
SIALT1375-100GEA 265 EUR
SIALT1375-100G SIALT1375-1KG SIALT1375-25G SIALT1375-500G SIALT1375-5KG
TES (2-(tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid), Sigma-Aldrich®
TES (2-(Tris(hydroxymethyl)methylamino)ethane-1-sulphonic acid)
Formula: C₆H₁₅NO₆S
Storage Temperature: Ambient
MDL Number: MFCD00007532
CAS Number: 7365-44-8
EINECS: 230-906-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR