Thiamine hydrochloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride

M-Clarity™ quality level = MQ200

T4625-250G T4625-5G T4625-100G T4625-10G T4625-1KG T4625-25G
SIALT4625-250GEA 283 EUR
SIALT4625-250G SIALT4625-5G SIALT4625-100G SIALT4625-10G SIALT4625-1KG SIALT4625-25G
Thiamine hydrochloride, Sigma-Aldrich®
Thiamine hydrochloride
Formula: C₁₂H₁₈Cl₂N₄OS
Melting Pt: 252 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR