Triethanolamine hydrochloride, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

T9534-100G T9534-1KG T9534-250G T9534-500G
SIALT9534-100GEA 177 EUR
SIALT9534-100G SIALT9534-1KG SIALT9534-250G SIALT9534-500G
Triethanolamine hydrochloride, Sigma-Aldrich®
Triethanolamine hydrochloride
Formula: C₆H₁₆ClNO₃
MW: 185.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012596
CAS Number: 637-39-8
EINECS: 211-284-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR