4-Methoxybenzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ300

W209902-10KG-K W209902-1KG-K W209902-25KG-K
SIALW209902-10KG-KEA 596 EUR
SIALW209902-10KG-K SIALW209902-1KG-K SIALW209902-25KG-K
4-Methoxybenzyl alcohol, Sigma-Aldrich®
4-Methoxybenzyl alcohol
Formula: C₈H₁₀O₂
Boiling Pt: 145 °C (1013 hPa)
Melting Pt: 20…24 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004653
CAS Number: 105-13-5
EINECS: 203-273-6
Merck Index: 13,00608

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR