Ethyl acetate, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Acetic acid ethyl ester

MQ300

W241407-10KG-K W241407-1KG-K W241407-20KG-K
SIALW241407-10KG-KEA 204 EUR
SIALW241407-10KG-K SIALW241407-1KG-K SIALW241407-20KG-K
Ethyl acetate, Sigma-Aldrich®
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR