Lauric acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Dodecanoic acid, Dodecylic acid

M-Clarity™ quality level = MQ300

W261408-10KG-K W261408-1KG-K W261408-25KG-K W261408-5KG-K
SIALW261408-10KG-KEA 230 EUR
SIALW261408-10KG-K SIALW261408-1KG-K SIALW261408-25KG-K SIALW261408-5KG-K
Lauric acid, Sigma-Aldrich®
Lauric acid
Formula: C₁₂H₂₄O₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 42…45 °C
Density: 0.875 g/cm³ (20 °C)
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00002736
CAS Number: 143-07-7
EINECS: 205-582-1
Merck Index: 13,05400

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR