2-Methylpyrazine, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

W330906-100G-K W330906-1KG-K W330906-5KG-K
SIALW330906-100G-KEA 101 EUR
SIALW330906-100G-K SIALW330906-1KG-K SIALW330906-5KG-K
2-Methylpyrazine, Sigma-Aldrich®
2-Methylpyrazine
Formula: C₅H₆N₂
Boiling Pt: 135 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.03 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006142
CAS Number: 109-08-0
EINECS: 203-645-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR