(±)-3-Methyl-2-butanol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: DL-3-Methyl-2-butanol

M-Clarity™ quality level = MQ300

W370304-100G W370304-1KG W370304-SAMPLE
SIALW370304-100GEA 106 EUR
SIALW370304-100G SIALW370304-1KG SIALW370304-SAMPLE
(±)-3-Methyl-2-butanol, Sigma-Aldrich®
(±)-3-Methyl-2-butanol
Formula: C₅H₁₂O
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00004527
CAS Number: 598-75-4
EINECS: 209-950-2
UN: 1105
ADR: 3,III
Merck Index: 12,06110

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR