TRITON™ X-305 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

X305-500ML X305-1GA
SIALX305-500MLEA 94.1 EUR
SIALX305-500ML SIALX305-1GA
TRITON™ X-305 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
TRITON™ X-305 (Polyethylene glycol tert-octylphenyl ether)
Storage Temperature: Ambient
CAS Number: 9002-93-1
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR