Ship propeller roughness comparator scales
Supplier: Industrial Physics
LD2041 LD2042
SIMELD2041EA 455 EUR
SIMELD2041 SIMELD2042
Ship propeller roughness comparator scales
Coating Testers
These roughness scales are designed to estimate the surface roughness, at touch or sight, specifically developed for the profiles related to the condition of ships propellers over life.
Ordering information: Each roughness scale includes six specimens.
Delivery information: Supplied with guidance manual and recommended limits instructions.