Stiffness tester, Taber®
Supplier: Industrial Physics
TB2200 TB2205
SIMETB2200EA 0 EUR
SIMETB2200 SIMETB2205
Stiffness tester, Taber®
Coating Testers
This stiffness tester is used to assess material stiffness and material resiliency for materials such as paper products, cardboard, plastics, textiles, metals, felts, tubing, wire, rubber and other sheet materials can be evaluated.
  • Accurate test measurements for specimens
  • Thickness of 0,10 mm to 5,56 mm with adjustable clamping system
  • Right hand roller with a ratchet stop mechanism
  • No interoperator error caused by over or under tightening
  • Travel on the right hand assembly automatically stops when the roller contacts the specimen
Bidirectional pendulum-More Product Information