Sample tubes
Supplier: Simport Scientific
C572-1 C572-2
SIMPC572-1EA 428 EUR
SIMPC572-1 SIMPC572-2
Sample tubes
Sample Containers General Purpose Containers
These tubes are chemically resistant and shatterproof, they are supplied with a leakproof screw cap, particularly important when transporting hazardous material.
  • Tube made of PP
  • Cap made of PE
  • Self-standing base with graduation
  • Leakproof screw cap
  • Available sterile and non-sterile
  • Functional temperature range: –90 to +121 °C
Conical bottom tubes with self-standing base graduated from 10 to 30 ml and 10 to 50 ml respectively in 5 ml increments. Ideal for transport and storage of urine, sputum and most liquids or particulate samples. Molded-inMore Product Information