Deep well plates, 96 well, Bioblock™
Supplier: Simport Scientific
T110-3 T110-2
SIMPT110-3EA 361 EUR
SIMPT110-3 SIMPT110-2
Deep well plates, 96 well, Bioblock™
Microplates Deep Well Microplates
All BioBlock™ Deepwell Plates are ideal for high volume, high throughput applications. The high-performance plates are chemical-resistant and designed for automated robotic handling.
  • Footprint conforms to SBS/ANSI standards
  • Great chemical stability
  • Free of RNase, DNase, Pyrogen, and DNA
  • Maximum sustainable centrifugal force up to 4000 g
  • The height difference from center to angle is less than 0,4 mm