Magnesium sulphate, anhydrous ≥99.0% USP

Supplier: Spectrum Chemical

Synonyms: MgSO4, Magnesium sulphate anhydrous

M1354-125GM
SPCMM1354-125GMEA 187 EUR
SPCMM1354-125GM
Magnesium sulphate, anhydrous ≥99.0% USP
Magnesium sulphate

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: MgSO₄
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 100.5%
pH (In Solution 1 in 20) 5.0 - 9.2
Loss On Drying max. 2 %
Chloride (Cl) max. 0.014 %
Iron max. 20 Ug/G


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR